2-Bromo-5-iodo-N-methoxy-N-methylbenzamide ≥95%

Supplier: AOBChem USA

Warning

65137-500MG 65137-1G 65137-5G
77332-444EA 328.57 USD
77332-444 77356-772 77380-716
2-Bromo-5-iodo-N-methoxy-N-methylbenzamide ≥95%
2-Bromo-5-iodo-N-methoxy-N-methylbenzamide

Precaución: for Research Use Only

Formula: C₉H₉BrINO₂
Storage Temperature: Ambient
MDL Number: MFCD34762117
CAS Number: 1699251-29-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR