4-Fluoro-3-(methylthio)-1-naphthoic acid ≥97%

Supplier: AOBChem USA

Warning

65124-500MG 65124-1G 65124-5G 65124-25G
77332-438EA 258.53 USD
77332-438 77356-764 77368-482 77389-738
4-Fluoro-3-(methylthio)-1-naphthoic acid ≥97%
4-Fluoro-3-(methylthio)-1-naphthoic acid

Precaución: for Research Use Only

Formula: C₁₂H₉FO₂S
MW: 236.2620 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762104
CAS Number: 2807455-60-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR