2-Methoxy-4'-methyl-5-(trifluoromethyl)-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

65105-500MG 65105-1G 65105-5G 65105-25G
77332-432EA 179.22 USD
77332-432 77346-742 77368-474 77389-732
2-Methoxy-4'-methyl-5-(trifluoromethyl)-1,1'-biphenyl ≥97%
2-Methoxy-4'-methyl-5-(trifluoromethyl)-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₅H₁₃F₃O
MW: 266.2583 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762099
CAS Number: 2807455-38-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR