4'-Fluoro-2-methoxy-5-(trifluoromethyl)-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

65103-500MG 65103-1G 65103-5G 65103-25G
77332-430EA 179.22 USD
77332-430 77346-740 77368-472 77389-730
4'-Fluoro-2-methoxy-5-(trifluoromethyl)-1,1'-biphenyl ≥97%
4'-Fluoro-2-methoxy-5-(trifluoromethyl)-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₀F₄O
MW: 270.2222 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14700951
CAS Number: 1214378-84-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR