2'-Methoxy-5'-(trifluoromethyl)-[1,1'-biphenyl]-4-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

65102-500MG 65102-1G 65102-5G 65102-25G
77332-428EA 147.29 USD
77332-428 77345-242 77368-470 77389-728
2'-Methoxy-5'-(trifluoromethyl)-[1,1'-biphenyl]-4-carbaldehyde ≥97%
2'-Methoxy-5'-(trifluoromethyl)-[1,1'-biphenyl]-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₅H₁₁F₃O₂
MW: 280.2418 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762098
CAS Number: 2807441-37-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR