2-Methoxy-3'-methyl-5-(trifluoromethyl)-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

65101-500MG 65101-1G 65101-5G 65101-25G
77332-426EA 136.99 USD
77332-426 77344-618 77368-468 77389-726
2-Methoxy-3'-methyl-5-(trifluoromethyl)-1,1'-biphenyl ≥95%
2-Methoxy-3'-methyl-5-(trifluoromethyl)-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₅H₁₃F₃O
MW: 266.2583 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762097
CAS Number: 2807448-65-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR