2-Methoxy-2'-methyl-5-(trifluoromethyl)-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

65100-500MG 65100-1G 65100-5G
77332-424EA 345.05 USD
77332-424 77356-760 77380-706
2-Methoxy-2'-methyl-5-(trifluoromethyl)-1,1'-biphenyl ≥97%
2-Methoxy-2'-methyl-5-(trifluoromethyl)-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₅H₁₃F₃O
MW: 266.2583 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762096
CAS Number: 2807443-31-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR