(5-Fluoro-2-methoxy-4-methylphenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

65098-500MG 65098-1G 65098-5G
77340-230EA 848.72 USD
77340-230 77356-758 77380-704
(5-Fluoro-2-methoxy-4-methylphenyl)boronic acid ≥97%
(5-Fluoro-2-methoxy-4-methylphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₁₀BFO₃
Storage Temperature: Ambient
MDL Number: MFCD23712918
CAS Number: 2093108-59-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR