5-Fluoro-2-methoxy-4-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

65095-500MG 65095-1G 65095-5G
77332-422EA 326.51 USD
77332-422 77356-756 77380-702
5-Fluoro-2-methoxy-4-methylbenzoic acid ≥97%
5-Fluoro-2-methoxy-4-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₉H₉FO₃
Storage Temperature: Ambient
MDL Number: MFCD23712094
CAS Number: 1427395-52-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR