1-(4-Chloro-2-methoxyphenyl)-2,2-difluoroethanone ≥97%

Supplier: AOBChem USA

Warning

65085-500MG 65085-1G 65085-5G 65085-25G
77332-420EA 178.19 USD
77332-420 77346-616 77368-466 77389-724
1-(4-Chloro-2-methoxyphenyl)-2,2-difluoroethanone ≥97%
1-(4-Chloro-2-methoxyphenyl)-2,2-difluoroethanone

Precaución: for Research Use Only

Formula: C₉H₇ClF₂O₂
MW: 220.6004 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30717139
CAS Number: 2090714-15-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR