3-Chloro-5-methyl-4-propoxybenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

65073-500MG 65073-1G 65073-5G
77337-824EA 448.05 USD
77337-824 77356-754 77380-700
3-Chloro-5-methyl-4-propoxybenzaldehyde ≥95%
3-Chloro-5-methyl-4-propoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₃ClO₂
MW: 212.6727 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06246361
CAS Number: 883517-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR