3-Chloro-5-methyl-4-propoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

65071-500MG 65071-1G 65071-25G 65071-5G
77332-418EA 285.31 USD
77332-418 77356-752 77389-722 77380-698
3-Chloro-5-methyl-4-propoxybenzoic acid ≥97%
3-Chloro-5-methyl-4-propoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₃ClO₃
MW: 228.6721 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762088
CAS Number: 1608148-19-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR