5-Fluoro-2-methoxy-4-methylbenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

65063-500MG 65063-1G
77340-452EA 1111.37 USD
77340-452 77356-750
5-Fluoro-2-methoxy-4-methylbenzaldehyde ≥95%
5-Fluoro-2-methoxy-4-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉FO₂
Storage Temperature: Ambient
MDL Number: MFCD06247546
CAS Number: 196519-56-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR