2-Ethoxy-5-fluoro-4-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

65057-500MG 65057-1G 65057-5G
77340-148EA 795.16 USD
77340-148 77356-748 77380-696
2-Ethoxy-5-fluoro-4-methylbenzoic acid ≥97%
2-Ethoxy-5-fluoro-4-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₃
MW: 198.1909 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762082
CAS Number: 2091890-85-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR