1-Bromo-2-ethoxy-5-fluoro-4-methylbenzene ≥97%
Supplier: AOBChem USA
|
Warning
|
65056-500MG
65056-1G
65056-5G
77337-942EA
453.2
USD
77337-942
77356-746
77380-694
1-Bromo-2-ethoxy-5-fluoro-4-methylbenzene ≥97%
1-Bromo-2-ethoxy-5-fluoro-4-methylbenzene
Precaución: for Research Use Only
Formula:
C₉H₁₀BrFO MW: 233.0775 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34762081 CAS Number: 2386042-97-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...