1-Bromo-5-fluoro-2-isopropoxy-4-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

65053-500MG 65053-1G 65053-5G
77337-964EA 455.26 USD
77337-964 77356-744 77380-692
1-Bromo-5-fluoro-2-isopropoxy-4-methylbenzene ≥97%
1-Bromo-5-fluoro-2-isopropoxy-4-methylbenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrFO
MW: 247.1040 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762079
CAS Number: 2807470-79-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR