3-Acetyl-5-ethoxy-2-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

65043-500MG 65043-1G 65043-5G 65043-25G
77332-414EA 233.81 USD
77332-414 77356-742 77368-462 77389-720
3-Acetyl-5-ethoxy-2-fluorobenzaldehyde ≥97%
3-Acetyl-5-ethoxy-2-fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₁FO₃
Storage Temperature: Ambient
MDL Number: MFCD34762072
CAS Number: 2385444-08-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR