4-Ethoxy-2-ethylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

65035-500MG 65035-1G 65035-5G
77337-730EA 438.78 USD
77337-730 77356-734 77380-684
4-Ethoxy-2-ethylbenzaldehyde ≥97%
4-Ethoxy-2-ethylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₂
MW: 178.2276 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06247339
CAS Number: 883536-89-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR