2-Ethyl-4-isopropoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

65031-500MG 65031-1G 65031-5G
77339-160EA 555.17 USD
77339-160 77356-732 77380-682
2-Ethyl-4-isopropoxybenzoic acid ≥97%
2-Ethyl-4-isopropoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₃
MW: 208.2536 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19309501
CAS Number: 882993-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR