2-Ethyl-4-isopropoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

65028-500MG 65028-1G 65028-5G
77339-960EA 693.19 USD
77339-960 77356-728 77380-680
2-Ethyl-4-isopropoxybenzaldehyde ≥97%
2-Ethyl-4-isopropoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₂
MW: 192.2542 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29121060
CAS Number: 1289168-88-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR