4-Fluoro-3-isobutoxy-5-(methylthio)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

65024-500MG 65024-1G 65024-5G
77335-888EA 367.71 USD
77335-888 77356-726 77380-678
4-Fluoro-3-isobutoxy-5-(methylthio)benzoic acid ≥97%
4-Fluoro-3-isobutoxy-5-(methylthio)benzoic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₅FO₃S
MW: 258.3091 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762064
CAS Number: 2807456-46-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR