2-Bromo-4-ethoxy-5-hydroxybenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

65021-500MG 65021-1G 65021-5G
77332-406EA 333.72 USD
77332-406 77356-722 77380-674
2-Bromo-4-ethoxy-5-hydroxybenzaldehyde ≥95%
2-Bromo-4-ethoxy-5-hydroxybenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉BrO₃
MW: 245.0699 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762061
CAS Number: 420800-56-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR