(3-Bromo-2-propoxyphenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

65017-500MG 65017-1G 65017-5G
77332-404EA 347.11 USD
77332-404 77356-718 77380-670
(3-Bromo-2-propoxyphenyl)(methyl)sulfane ≥95%
(3-Bromo-2-propoxyphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₀H₁₃BrOS
MW: 261.1786 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762060
CAS Number: 2807447-98-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR