1-Bromo-3-iodo-2-propoxybenzene ≥97%
Supplier: AOBChem USA
|
Warning
|
65016-500MG
65016-1G
65016-5G
77332-402EA
340.93
USD
77332-402
77356-716
77380-668
1-Bromo-3-iodo-2-propoxybenzene ≥97%
1-Bromo-3-iodo-2-propoxybenzene
Precaución: for Research Use Only
Formula:
C₉H₁₀BrIO MW: 340.9835 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34762059 CAS Number: 2807471-01-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...