4-Chloro-3,4'-dimethoxy-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

65012-500MG 65012-1G 65012-5G
77332-400EA 347.11 USD
77332-400 77356-714 77380-666
4-Chloro-3,4'-dimethoxy-1,1'-biphenyl ≥97%
4-Chloro-3,4'-dimethoxy-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₃ClO₂
MW: 248.7048 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762058
CAS Number: 74447-82-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR