4-Chloro-3-methoxy-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

65010-500MG 65010-1G 65010-5G
77332-396EA 347.11 USD
77332-396 77356-710 77380-662
4-Chloro-3-methoxy-1,1'-biphenyl ≥95%
4-Chloro-3-methoxy-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₁₁ClO
MW: 218.6788 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14701649
CAS Number: 74447-81-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR