2-Bromo-6-(methylthio)phenol ≥97%

Supplier: AOBChem USA

Warning

65007-500MG 65007-1G 65007-25G 65007-5G
77332-394EA 288.4 USD
77332-394 77356-708 77389-712 77380-660
2-Bromo-6-(methylthio)phenol ≥97%
2-Bromo-6-(methylthio)phenol

Precaución: for Research Use Only

Formula: C₇H₇BrOS
MW: 219.0988 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16999071
CAS Number: 107724-67-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR