3-(4-Chloro-3-fluorophenyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

65004-500MG 65004-1G 65004-5G
77340-040EA 722.03 USD
77340-040 77356-706 77380-658
3-(4-Chloro-3-fluorophenyl)pyridine ≥97%
3-(4-Chloro-3-fluorophenyl)pyridine

Precaución: for Research Use Only

Formula: C₁₁H₇ClFN
MW: 207.6313 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30752828
CAS Number: 2137736-85-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR