4-(4-Chloro-3-fluorophenyl)pyridine ≥95%

Supplier: AOBChem USA

Warning

65003-500MG 65003-1G 65003-5G 65003-25G
77332-392EA 116.39 USD
77332-392 77343-446 77368-456 77380-656
4-(4-Chloro-3-fluorophenyl)pyridine ≥95%
4-(4-Chloro-3-fluorophenyl)pyridine

Precaución: for Research Use Only

Formula: C₁₁H₇ClFN
MW: 207.6313 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762056
CAS Number: 2344704-57-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR