4-Chloro-3,4'-difluoro-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

65002-500MG 65002-1G 65002-5G 65002-25G
77332-390EA 116.39 USD
77332-390 77343-500 77368-454 77380-654
4-Chloro-3,4'-difluoro-1,1'-biphenyl ≥95%
4-Chloro-3,4'-difluoro-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₂H₇ClF₂
Storage Temperature: Ambient
MDL Number: MFCD34762055
CAS Number: 2386082-36-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR