4-Chloro-3-fluoro-4'-methyl-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

65000-500MG 65000-1G 65000-5G 65000-25G
77332-388EA 134.93 USD
77332-388 77344-478 77368-452 77389-710
4-Chloro-3-fluoro-4'-methyl-1,1'-biphenyl ≥95%
4-Chloro-3-fluoro-4'-methyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₁₀ClF
MW: 220.6699 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06802330
CAS Number: 524018-93-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR