2-Chloro-4,4'-dimethoxy-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

64996-500MG 64996-1G 64996-5G
77338-164EA 466.59 USD
77338-164 77356-704 77380-652
2-Chloro-4,4'-dimethoxy-1,1'-biphenyl ≥97%
2-Chloro-4,4'-dimethoxy-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₃ClO₂
Storage Temperature: Ambient
MDL Number: MFCD34762052
CAS Number: 74447-86-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR