2-Chloro-4-methoxy-4'-methyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

64994-500MG 64994-1G 64994-5G
77337-208EA 408.91 USD
77337-208 77356-702 77380-650
2-Chloro-4-methoxy-4'-methyl-1,1'-biphenyl ≥97%
2-Chloro-4-methoxy-4'-methyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₃ClO
Storage Temperature: Ambient
MDL Number: MFCD34762051
CAS Number: 2807471-51-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR