2-Bromo-1-iodo-4-(2-methoxyethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

64992-500MG 64992-1G 64992-5G 64992-25G
77332-384EA 202.91 USD
77332-384 77356-700 77368-448 77389-706
2-Bromo-1-iodo-4-(2-methoxyethoxy)benzene ≥97%
2-Bromo-1-iodo-4-(2-methoxyethoxy)benzene

Precaución: for Research Use Only

Formula: C₉H₁₀BrIO₂
MW: 356.9829 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762049
CAS Number: 1862522-39-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR