4-(Benzyloxy)-2-bromobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

64982-500MG 64982-1G 64982-5G
77332-382EA 307.97 USD
77332-382 77356-698 77380-648
4-(Benzyloxy)-2-bromobenzoic acid ≥97%
4-(Benzyloxy)-2-bromobenzoic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₁BrO₃
MW: 307.1393 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762044
CAS Number: 1874170-93-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR