2-Bromo-4-isopropoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

64976-500MG 64976-1G 64976-5G 64976-25G
77332-378EA 257.5 USD
77332-378 77356-694 77368-444 77389-702
2-Bromo-4-isopropoxybenzoic acid ≥97%
2-Bromo-4-isopropoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrO₃
MW: 259.0965 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24368751
CAS Number: 1369882-92-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR