3-Bromo-5-methyl-2-(2,2,2-trifluoroethoxy)pyridine ≥97%

Supplier: AOBChem USA

Warning

64973-500MG 64973-1G 64973-5G 64973-25G
77332-376EA 126.69 USD
77332-376 77344-190 77368-442 77380-646
3-Bromo-5-methyl-2-(2,2,2-trifluoroethoxy)pyridine ≥97%
3-Bromo-5-methyl-2-(2,2,2-trifluoroethoxy)pyridine

Precaución: for Research Use Only

Formula: C₈H₇BrF₃NO
MW: 270.0464 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29765137
CAS Number: 760207-90-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR