1-(2-(Benzyloxy)-4-chlorophenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

64968-500MG 64968-1G 64968-5G 64968-25G
77332-370EA 95.79 USD
77332-370 77342-582 77368-436 77380-644
1-(2-(Benzyloxy)-4-chlorophenyl)ethanone ≥97%
1-(2-(Benzyloxy)-4-chlorophenyl)ethanone

Precaución: for Research Use Only

Formula: C₁₅H₁₃ClO₂
MW: 260.7155 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15528683
CAS Number: 76292-53-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR