6-(Cyclopropylmethoxy)-5-methylnicotinic acid ≥97%

Supplier: AOBChem USA

Warning

64956-500MG 64956-1G 64956-5G
77338-162EA 466.59 USD
77338-162 77356-690 77380-640
6-(Cyclopropylmethoxy)-5-methylnicotinic acid ≥97%
6-(Cyclopropylmethoxy)-5-methylnicotinic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₃NO₃
Storage Temperature: Ambient
MDL Number: MFCD28963287
CAS Number: 1476027-21-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR