3-Fluoro-2,5-dimethylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

64945-500MG 64945-1G 64945-5G
77337-792EA 443.93 USD
77337-792 77356-686 77380-636
3-Fluoro-2,5-dimethylbenzoic acid ≥97%
3-Fluoro-2,5-dimethylbenzoic acid

Precaución: for Research Use Only

Formula: C₉H₉FO₂
MW: 168.1649 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23712081
CAS Number: 1427433-77-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR