Methyl 5-chloro-6-methoxypicolinate ≥95%

Supplier: AOBChem USA

Warning

64927-500MG 64927-1G 64927-5G 64927-25G
77332-362EA 175.1 USD
77332-362 77346-542 77368-432 77389-690
Methyl 5-chloro-6-methoxypicolinate ≥95%
Methyl 5-chloro-6-methoxypicolinate

Precaución: for Research Use Only

Formula: C₈H₈ClNO₃
MW: 201.6070 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14698152
CAS Number: 1214386-12-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR