2,2,2-trifluoro-1-(2-fluoro-3-(trifluoromethyl)phenyl)ethanone ≥97%
Supplier: AOBChem USA
|
Warning
|
64897-500MG
64897-1G
64897-5G
77336-540EA
388.31
USD
77336-540
77356-678
77380-628
2,2,2-trifluoro-1-(2-fluoro-3-(trifluoromethyl)phenyl)ethanone ≥97%
2,2,2-trifluoro-1-(2-fluoro-3-(trifluoromethyl)phenyl)ethanone
Precaución: for Research Use Only
Formula:
C₉H₃F₇O MW: 260.1083 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34762018 CAS Number: 1503058-48-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...