1-(3-Chloro-2,4-difluorophenyl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

64890-500MG 64890-1G 64890-5G
77336-538EA 388.31 USD
77336-538 77356-676 77380-626
1-(3-Chloro-2,4-difluorophenyl)-2,2,2-trifluoroethanone ≥95%
1-(3-Chloro-2,4-difluorophenyl)-2,2,2-trifluoroethanone

Precaución: for Research Use Only

Formula: C₈H₂ClF₅O
MW: 244.5458 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762015
CAS Number: 2029517-03-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR