1-Bromo-2-chloro-5-nitro-3-(trifluoromethyl)benzene ≥95%
Supplier: AOBChem USA
|
Warning
|
64888-500MG
64888-1G
64888-5G
64888-25G
77332-356EA
252.35
USD
77332-356
77356-672
77368-424
77389-684
1-Bromo-2-chloro-5-nitro-3-(trifluoromethyl)benzene ≥95%
1-Bromo-2-chloro-5-nitro-3-(trifluoromethyl)benzene
Precaución: for Research Use Only
Formula:
C₇H₂BrClF₃NO₂ MW: 304.4484 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11520116 CAS Number: 1096698-02-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...