3-Bromo-6-isopropoxy-2-methylaniline ≥97%

Supplier: AOBChem USA

Warning

64859-500MG 64859-1G 64859-5G
77337-580EA 424.36 USD
77337-580 77356-668 77380-622
3-Bromo-6-isopropoxy-2-methylaniline ≥97%
3-Bromo-6-isopropoxy-2-methylaniline

Precaución: for Research Use Only

Formula: C₁₀H₁₄BrNO
MW: 244.1282 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30096940
CAS Number: 1865086-15-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR