1-Bromo-4-isopropoxy-2-methyl-3-nitrobenzene ≥97%

Supplier: AOBChem USA

Warning

64858-500MG 64858-1G 64858-5G
77335-714EA 363.59 USD
77335-714 77356-666 77380-620
1-Bromo-4-isopropoxy-2-methyl-3-nitrobenzene ≥97%
1-Bromo-4-isopropoxy-2-methyl-3-nitrobenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrNO₃
MW: 274.1111 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30729342
CAS Number: 2169195-42-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR