3-Bromo-6-ethoxy-2-methylaniline ≥97%

Supplier: AOBChem USA

Warning

64851-500MG 64851-1G 64851-5G
77337-578EA 424.36 USD
77337-578 77356-664 77380-618
3-Bromo-6-ethoxy-2-methylaniline ≥97%
3-Bromo-6-ethoxy-2-methylaniline

Precaución: for Research Use Only

Formula: C₉H₁₂BrNO
Storage Temperature: Ambient
MDL Number: MFCD30729353
CAS Number: 1866137-63-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR