1-Bromo-3-iodo-4-methoxy-2-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

64849-500MG 64849-1G 64849-5G
77336-536EA 388.31 USD
77336-536 77356-660 77380-616
1-Bromo-3-iodo-4-methoxy-2-methylbenzene ≥97%
1-Bromo-3-iodo-4-methoxy-2-methylbenzene

Precaución: for Research Use Only

Formula: C₈H₈BrIO
Storage Temperature: Ambient
MDL Number: MFCD34761991
CAS Number: 2383765-92-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR