(2-Chloropyridin-3-yl)(thiomorpholino)methanone ≥97%

Supplier: AOBChem USA

Warning

64848-1G 64848-5G 64848-25G
77340-796EA 60.77 USD
77340-796 77368-416 77380-614
(2-Chloropyridin-3-yl)(thiomorpholino)methanone ≥97%
(2-Chloropyridin-3-yl)(thiomorpholino)methanone

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClN₂OS
MW: 242.7251 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11172209
CAS Number: 1042653-50-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR