1-(3-Chloro-5-fluorophenyl)piperidine ≥97%

Supplier: AOBChem USA

Warning

64841-500MG 64841-1G 64841-5G 64841-25G
77332-346EA 108.15 USD
77332-346 77343-194 77368-412 77380-610
1-(3-Chloro-5-fluorophenyl)piperidine ≥97%
1-(3-Chloro-5-fluorophenyl)piperidine

Precaución: for Research Use Only

Formula: C₁₁H₁₃ClFN
MW: 213.6790 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761989
CAS Number: 2901103-57-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR